Merck
  • Impaired Th1 immunity in ovarian cancer patients is mediated by TNFR2+ Tregs within the tumor microenvironment.
Impaired Th1 immunity in ovarian cancer patients is mediated by TNFR2+ Tregs within the tumor microenvironment.
Clinical immunology (Orlando, Fla.) (2013-08-21)
Chindu Govindaraj, Karen Scalzo-Inguanti, Mutsa Madondo, Julene Hallo, Katie Flanagan, Michael Quinn, Magdalena Plebanski
ABSTRACT
Ovarian cancer is a prevalent gynecological malignancy with potent immune-suppression capabilities; regulatory T cells (Tregs) are significant contributors to this immune-suppression. As ovarian cancer patients present with high levels of TNF and Tregs expressing TNFR2 are associated with maximal suppressive capacity, we investigated TNFR2+ Tregs within these patients. Indeed, TNFR2+ Tregs from tumor-associated ascites were the most potent suppressor T cell fraction. They were abundantly present within the ascites and more suppressive than peripheral blood TNFR2+ Tregs in patients. The increased suppressive capacity can be explained by a distinct cell surface expression profile, which includes high levels of CD39, CD73, TGF-β and GARP. Additionally, CD73 expression level on TNFR2+ Tregs was inversely correlated with IFN-γ production by effector T cells. This Treg fraction can be selectively recruited into the ascites from the peripheral blood of patients. Targeting TNFR2+ Tregs may offer new approaches to enhance the poor survival rates of ovarian cancer.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder