Merck
  • Assessment of the rate of long-term complete remission off therapy in patients with pemphigus treated with different regimens including medium- and high-dose corticosteroids.
Assessment of the rate of long-term complete remission off therapy in patients with pemphigus treated with different regimens including medium- and high-dose corticosteroids.
Journal of the American Academy of Dermatology (2013-07-16)
Naif Almugairen, Valerie Hospital, Christophe Bedane, Sophie Duvert-Lehembre, Damien Picard, Anne-Fleur Tronquoy, Estelle Houivet, Michel D'incan, Pascal Joly
ABSTRACT
Few studies have evaluated pemphigus treatments according to the definitions of the consensus statement. Prognostic factors for complete remission off therapy (CRoffT) remain unknown. We sought to assess the rate of CRoffT in patients with pemphigus treated with different regimens. In all, 134 patients with pemphigus were included in a retrospective, multicenter study. Primary end point was the rate of CRoffT. Prognostic factors for CRoffT were determined using univariate and multivariate analyses. Eighty patients with pemphigus vulgaris, 47 with pemphigus foliaceus, and 7 with paraneoplastic pemphigus were included. Mean age was 60 ± 18 years. Patients were treated either with medium (≤0.5 mg/kg/d) (n = 32) or high (≥1 mg/kg/d) (n = 59) doses of prednisone, or without systemic corticosteroids (n = 43). Mean follow-up was 77 ± 64 months. In all, 68 patients (50.7%) achieved CRoffT (95% confidence interval 42.3%-59.2%) after a mean treatment duration of 36 ± 39 months, including 47 of 80 patients with pemphigus vulgaris (58.7%) and 21 of 47 with pemphigus foliaceus (44.7%). Main prognostic factors for CRoffT were initial mucosal involvement (hazard ratio 2.2; 95% confidence interval 1.05-4.58; P = .036) and younger age (<61 years) (hazard ratio 2.5; 95% confidence interval 1.18-5.12; P = .0167). The rate of long-lasting CRoffT was 44%, with a mean follow-up after treatment withdrawal of 59 ± 50 months. This was a retrospective study. The rate of CRoffT was 51%. Patients with pemphigus vulgaris were more likely to achieve CRoffT than those with pemphigus foliaceus.
Product Description
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard