Merck
  • Expression levels of neuroimmune biomarkers in hypothalamus of allergic mice after phthalate exposure.
Expression levels of neuroimmune biomarkers in hypothalamus of allergic mice after phthalate exposure.
Journal of applied toxicology : JAT (2012-11-14)
Tin-Tin Win-Shwe, Rie Yanagisawa, Eiko Koike, Hiroshi Nitta, Hirohisa Takano
ABSTRACT
Previously, we demonstrated that maternal exposure to phthalates enhances atopic dermatitis in male mouse offspring. However, whether phthalate exposure affects neuroimmune biomarkers in allergic mice has not yet been studied. Di-(2-ethylhexyl) phthalate (DEHP) and di-isononyl phthalate (DINP) are environmental chemicals that are commonly used as plasticizers. This study was designed to investigate the expression levels of neuroimmune biomarkers in the hypothalamus of a murine model of allergic asthma after phthalate exposure throughout juvenility until adulthood. Six-week-old C3H/HeJ Jcl male mice were treated with DEHP or DINP (0, 0.02, 0.4 or 8 nmol per body per week) and ovalbumin (OVA; 1 µg per body per 2 weeks) for 7 weeks intratracheally. On the day after the completion of the phthalate and OVA treatment, the hypothalamus from each mouse was collected, and the mRNA expression levels of neuroimmune biomarkers were examined using a real-time RT-PCR analysis. The mRNA expression levels of the proinflammatory cytokines interleukin (IL)-1β and tumor necrosis factor (TNF)-α, the chemokine CCL3, the transcription factor nuclear factor (NF)-κB, the oxidative stress marker heme-oxygenase (HO)1, a nerve growth factor, and the microglia marker Iba1 were remarkably up-regulated in the hypothalami of mice treated with 8 nmol of DEHP in the presence of the allergen. However, no significant changes were observed, except for reductions in the TNF-α and CCL2 mRNA levels, in mice exposed to DINP combined with the allergen. This study is the first report to show that high-dose DEHP exposure throughout juvenility until adulthood may induce neuroinflammation by modulating neuroimmune biomarkers in the hypothalami of allergic mice.
Product Description
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%