Merck
  • KOMET: an unblinded, randomised, two parallel-group, stratified trial comparing the effectiveness of levetiracetam with controlled-release carbamazepine and extended-release sodium valproate as monotherapy in patients with newly diagnosed epilepsy.
KOMET: an unblinded, randomised, two parallel-group, stratified trial comparing the effectiveness of levetiracetam with controlled-release carbamazepine and extended-release sodium valproate as monotherapy in patients with newly diagnosed epilepsy.
Journal of neurology, neurosurgery, and psychiatry (2012-08-31)
Eugen Trinka, Anthony G Marson, Wim Van Paesschen, Reetta Kälviäinen, Jacqueline Marovac, Benjamin Duncan, Sonja Buyle, Yngve Hallström, Petr Hon, Gian Carlo Muscas, Mark Newton, Heinz-Joachim Meencke, Philip E Smith, Bernd Pohlmann-Eden
ABSTRACT
To compare the effectiveness of levetiracetam (LEV) with extended-release sodium valproate (VPA-ER) and controlled-release carbamazepine (CBZ-CR) as monotherapy in patients with newly diagnosed epilepsy. This unblinded, randomised, 52-week superiority trial (NCT00175903) recruited patients (≥16 years of age) with ≥2 unprovoked seizures in the previous 2 years and ≥1 in the previous 6 months. The physician chose VPA or CBZ as preferred standard treatment; each patient was randomised to standard treatment or LEV. The primary outcome was time to treatment withdrawal (LEV vs standard antiepileptic drugs (AEDs)). Analyses also compared LEV with VPA-ER, and LEV with CBZ-CR. 1688 patients (mean age 41 years; 44% female) were randomised to LEV (n=841) or standard AEDs (n=847). Time to treatment withdrawal was not significantly different between LEV and standard AEDs: HR (95% CI) 0.90 (0.74 to 1.08). Time to treatment withdrawal (HR (95% CI)) was 1.02 (0.74 to 1.41) for LEV/VPA-ER and 0.84 (0.66 to 1.07) for LEV/CBZ-CR. Time to first seizure (HR, 95% CI) was significantly longer for standard AEDs, 1.20 (1.03 to 1.39), being 1.19 (0.93 to 1.54) for LEV/VPA-ER and 1.20 (0.99 to 1.46) for LEV/CBZ-CR. Estimated 12-month seizure freedom rates from randomisation: 58.7% LEV versus 64.5% VPA-ER; 50.5% LEV versus 56.7% CBZ-CR. Similar proportions of patients within each stratum reported at least one adverse event: 66.1% LEV versus 62.0% VPA-ER; 73.4% LEV versus 72.5% CBZ-CR. LEV monotherapy was not superior to standard AEDs for the global outcome, namely time to treatment withdrawal, in patients with newly diagnosed focal or generalised seizures.
Product Description
Levetiracetam impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Piracetam
Supelco
Levetiracetam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Levetiracetam, European Pharmacopoeia (EP) Reference Standard