Merck
Protecting-group-free total synthesis of (+/-)-subincanadine F.
The Journal of organic chemistry (2009-09-26)
Pinhong Chen, Lidong Cao, Chaozhong Li
ABSTRACT
With chemoselective Dieckmann condensation as the key step, the protective-group-free total synthesis of (+/-)-subincanadine F was accomplished in 7 steps from the commercially available tryptamine in 33% overall yield.
Sigma-Aldrich
Methyl bromopyruvate, technical
Pricing and availability is not currently available.