Merck
  • Reactions of halobenzenes with methanol on the microporous solid acids hbeta, HZSM-5, and HSAPO-5: halogenation does not improve the hydrocarbon pool.
Reactions of halobenzenes with methanol on the microporous solid acids hbeta, HZSM-5, and HSAPO-5: halogenation does not improve the hydrocarbon pool.
Langmuir : the ACS journal of surfaces and colloids (2006-02-08)
David M Marcus, Weiguo Song, Saifudin M Abubakar, Emma Jani, Alain Sassi, James F Haw
ABSTRACT
The reactions of fluorobenzene, 3-fluorotoluene, and three isomers of difluorotoluene, chlorobenzene, and bromobenzene with excesses of methanol were investigated on the large-pore catalysts HBeta (*BEA) and HSAPO-5 (AFI), and on the medium-pore HZSM-5 (MFI). Flow reactor studies in pulse mode with GC-MS detection revealed that the fluorobenzene derivatives were readily methylated at, for example, 375 degrees C, but not even pentamethylfluorobenzene was obviously active as a reaction center for methanol-to-olefin (MTO) catalysis. Carbon-labeling studies revealed that small amounts of methylbenzenes were formed by defluorination, and these aromatic hydrocarbons seemed to account for the small yields of olefins (and their secondary reaction products) observed. Loss of one fluorine was also evident in the products for one of the difluorotoluene isomers. On HSAPO-5 the activity order for ring-methylation of halobenzenes was F > Cl > Br. On HZSM-5, chlorobenzene and especially bromobenzene lost halogen through a route forming halomethane. These largely negative results will nevertheless be useful in testing theoretical models of the detailed reaction steps in the hydrocarbon pool mechanism for MTO catalysis.
Sigma-Aldrich
3-Fluorotoluene, 99%
Pricing and availability is not currently available.