Merck
  • Final report on the safety assessment of amino nitrophenols as used in hair dyes.
Final report on the safety assessment of amino nitrophenols as used in hair dyes.
International journal of toxicology (2010-01-30)
Christina L Burnett, Wilma F Bergfeld, Donald V Belsito, Curtis D Klaassen, James G Marks, Ronald C Shank, Thomas J Slaga, Paul W Snyder, F Alan Andersen
ABSTRACT
2-Amino-3-nitrophenol, 2-amino-4-nitrophenol, 2-amino-5-nitrophenol, 4-amino-3-nitrophenol, 4-amino-2-nitrophenol, 2-amino-4-nitrophenol sulfate, 3-nitro-p-hydroxyethylaminophenol, and 4-hydroxypropylamino-3-nitrophenol are substituted aromatic compounds used as semipermanent (nonoxidative) hair colorants and as toners in permanent (oxidative) hair dye products. All ingredients in this group except 2-amino-4-nitrophenol sulfate, 2-amino-5-nitrophenol, and 4-amino-2-nitrophenol have reported uses in cosmetics at use concentrations from 2% to 9%. The available toxicity studies for these amino nitrophenol hair dyes did not suggest safety concerns except for the potential carcinogenicity and mutagenicity of 4-amino-2-nitrophenol. 2-Amino-3-nitrophenol, 2-amino-4-nitrophenol, 2-amino-4-nitrophenol sulfate, 2-amino-5-nitrophenol, 4-amino-3-nitrophenol, 3-nitro-p-hydroxyethylaminophenol, and 4-hydroxypropylamino-3-nitrophenol are safe as hair dye ingredients in the practices of use and concentration as described in this safety assessment, but the data are insufficient to make a safety determination for 4-amino-2-nitrophenol.
Sigma-Aldrich
4-Amino-2-nitrophenol, 97%
Sigma-Aldrich
2-Amino-5-nitrophenol
Sigma-Aldrich
2-Amino-3-nitrophenol, 98%