- Palladium-catalyzed synthesis of indene derivatives via propargylic carbonates with in situ generated organozinc compounds.
Palladium-catalyzed synthesis of indene derivatives via propargylic carbonates with in situ generated organozinc compounds.
Organic & biomolecular chemistry (2008-03-11)
Zheng-Hui Guan, Zhi-Hui Ren, Lian-Biao Zhao, Yong-Min Liang
PMID18327329
ABSTRACT
The palladium-catalyzed carboannulation and arylation reaction of propargylic carbonates with in situ generated organozinc compounds produced an important new class of indene derivatives. The reaction proceeded under mild conditions, and indene products were isolated in good to excellent yields.