Merck
  • Effect of all-trans retinoic acid on procoagulant and fibrinolytic activities of cultured blast cells from patients with acute promyelocytic leukemia.
Effect of all-trans retinoic acid on procoagulant and fibrinolytic activities of cultured blast cells from patients with acute promyelocytic leukemia.
Blood (1995-11-01)
V De Stefano, L Teofili, S Sica, S Mastrangelo, A Di Mario, S Rutella, P Salutari, C Rumi, G d'Onofrio, G Leone
ABSTRACT
The mechanisms underlying acute promyelocytic leukemia (APL) coagulopathy and its reversal by administration of all-trans retinoic acid (ATRA) have been investigated. Bone marrow promyelocytic blasts from nine patients with APL were cultured with or without ATRA 1 mumol/L. Cultured blasts (days 0, 3, 6, and 9) were washed, resuspended in phosphate buffer, lysed by freezing and thawing, and then assayed for procoagulant activity (PCA), elastase activity, tissue factor (TF) antigen, tissue-type plasminogen activator (t-PA) antigen and urokinase-type plasminogen activator (u-PA) antigen. PCA was determined by a recalcification assay. Elastase was measured by an amidolytic assay (S-2484). TF, t-PA, and u-PA antigens were measured by an enzyme-linked immunosorbent assay (ELISA). Malignant promyelocytes isolated from the patients had increased levels of PCA and TF as compared with the control polymorphonucleates, and low levels of elastase, t-PA, and u-PA; the patient blast PCA level was significantly related to the degree of hypofibrinogenemia. In this system, blast PCA depended on the tissue factor and was significantly correlated to the TF antigen values. In the cultures without ATRA, PCA, TF, and u-PA progressively increased, whereas elastase and t-PA levels remained essentially unchanged. In the presence of ATRA, all parameters (except u-PA) decreased during the culture time. Thus, a major role of the promyelocytic blast cell PCA in the pathogenesis of M3-related coagulopathy is suggested; the ATRA effect on coagulopathy seems mainly mediated by a downregulation of the PCA.
Sigma-Aldrich
Elastase from human leukocytes, lyophilized powder, ≥50 units/mg protein (Bradford)