Merck
  • Catalytic photooxidation of 4-methoxybenzyl alcohol with a flavin-zinc(II)-cyclen complex.
Catalytic photooxidation of 4-methoxybenzyl alcohol with a flavin-zinc(II)-cyclen complex.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-10-16)
Radek Cibulka, Rudolf Vasold, Burkhard König
ABSTRACT
Flavin-zinc(II)-cyclen 10 contains a covalently linked substrate binding site (zinc(II)-cyclen) and a chromophore unit (flavin). Upon irradiation, compound 10 effectively oxidizes 4-methoxybenzyl alcohol (11-OCH3) to the corresponding benzaldehyde both in water and in acetonitrile. In the presence of air, the reduced flavin 10-H2 is reoxidized, and so catalytic amounts of 10 are sufficient for alcohol conversion. The mechanism of oxidation is based on photoinduced electron transfer from the coordinated benzyl alcohol to the flavin chromophore. This intramolecular process provides a much higher photooxidation efficiency, with quantum yields 30 times those of the comparable intermolecular process with a flavin chromophore without a binding site. For the reaction in buffered aqueous solution a quantum yield of Phi = 0.4 is observed. The turnover number in acetonitrile is increased (up to 20) by high benzyl alcohol concentrations. The results show that the covalent combination of a chromophore and a suitable binding site may lead to photomediators more efficient than classical sensitizer molecules.
Sigma-Aldrich
4-Methoxybenzyl alcohol, 98%