Merck
  • A concise synthesis of all four possible benzo[4,5]furopyridines via palladium-mediated reactions.
A concise synthesis of all four possible benzo[4,5]furopyridines via palladium-mediated reactions.
Organic letters (2002-06-21)
Wen Song Yue, Jie Jack Li
ABSTRACT
[reaction: see text] By taking advantage of the alpha- and gamma-activation of chloropyridines as well as palladium-mediated reactions, all four possible benzo[4,5]furopyridine tricyclic heterocycles, benzo[4,5]furo[2,3-b]pyridine, benzo[4,5]furo[2,3-c]pyridine, benzo[4,5]furo[3,2-c]pyridine, and benzo[4,5]furo[3,2-b]pyridine, are efficiently synthesized from 2-chloro-3-iodopyridine, 3-chloro-4-stannylpyridine, 4-chloro-3-iodopyridine, and 2-chloro-3-hydroxypyridine, respectively.
Sigma-Aldrich
2-Chloro-3-hydroxypyridine, 98%