Merck
  • Enrichment of dibenzofuran utilizing bacteria with high co-metabolic potential towards dibenzodioxin and other anellated aromatics.
Enrichment of dibenzofuran utilizing bacteria with high co-metabolic potential towards dibenzodioxin and other anellated aromatics.
FEMS microbiology letters (1989-04-01)
V Strubel, H G Rast, W Fietz, H J Knackmuss, K H Engesser
ABSTRACT
Dibenzofuran degrading bacteria were enriched from various environmental sources. A mutualistic mixed culture of strain DPO 220 and strain DPO 230 was characterized. Strain DPO 220 alone showed limited growth with dibenzofuran as sole source of carbon and energy (td greater than or equal to 4.5 h). A labile degradation product, C12H10O5, and salicylate were isolated from the culture fluid. Salicylate was found to be a central intermediate of DBF-degradation. Strain DPO 220 co-metabolized a wide range of anellated aromatics as well as heteroaromatics. High rates of co-oxidation of dibenzodioxin demonstrate analogue-enrichment to be a powerful technique for selecting enzymatic activities for otherwise non-degradable substrates.
Sigma-Aldrich
2-Coumaranone, 97%