Merck
  • Facile O-deallylation of allyl ethers via S(N)2' reaction with tert-butyllithium
Facile O-deallylation of allyl ethers via S(N)2' reaction with tert-butyllithium
Organic letters (2000-05-18)
Bailey, England, Mealy, Thongsornkleeb, Teng
ABSTRACT
[reaction: see text] Allylic ethers are converted to the corresponding alcohol or phenol in virtually quantitative yield at temperatures below ambient simply by stirring a hydrocarbon solution of the ether with 1 molar equiv of tert-butyllithium. The reaction, which produces 4,4-dimethyl-1-pentene as a coproduct, most likely involves an S(N)2' attack of the organolithium on the allyl ether.
Sigma-Aldrich
4,4-Dimethyl-1-pentene, 99%