Merck
  • LPS regulates SOCS2 transcription in a type I interferon dependent autocrine-paracrine loop.
LPS regulates SOCS2 transcription in a type I interferon dependent autocrine-paracrine loop.
PloS one (2012-02-01)
Jin Hu, DaoHua Lou, Berit Carow, Malin E Winerdal, Martin Rottenberg, Ann-Charlotte Wikström, Gunnar Norstedt, Ola Winqvist
ABSTRACT
Recent studies suggest that SOCS2 is involved in the regulation of TLR signaling. In this study, we found that the expression of SOCS2 is regulated in human monocyte-derived DC by ligands stimulating TLR2, 3, 4, 5, 8 and 9 signaling. SOCS2 induction by LPS was dependent on the type I IFN regulated transcription factors IRF1 and IRF3 as shown by using silencing RNAs for IRFs. Blocking endogenous type I IFN signaling, by neutralizing antibodies to the receptor IFNAR2, abolished SOCS2 mRNA expression after TLR4 stimulation. Transcription factors STAT3, 5 and 6 displayed putative binding sites in the promoter regions of the human SOCS2 gene. Subsequent silencing experiments further supported that STAT3 and STAT5 are involved in LPS induced SOCS2 regulation. In mice we show that SOCS2 mRNA induction is 45% lower in bone marrow derived macrophages derived from MyD88(-/-) mice, and do not increase in BMMs from IRF3(-/-) mice after BCG infection. In conclusion, our results suggest that TLR4 signaling indirectly increases SOCS2 in late phase mainly via the production of endogenous type I IFN, and that subsequent IFN receptor signaling activates SOCS2 via STAT3 and STAT5.
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture