Merck
  • Effects of an agonist, allosteric modulator, and antagonist on guanosine-gamma-[35S]thiotriphosphate binding to liposomes with varying muscarinic receptor/Go protein stoichiometry.
Effects of an agonist, allosteric modulator, and antagonist on guanosine-gamma-[35S]thiotriphosphate binding to liposomes with varying muscarinic receptor/Go protein stoichiometry.
Molecular pharmacology (1998-11-06)
J Jakubík, T Haga, S Tucek
ABSTRACT
We investigated whether alcuronium, an allosteric modulator of muscarinic acetylcholine receptors, can induce receptor-mediated activation of Go proteins in liposomal membranes incorporating purified M2 receptors and Go proteins and whether its action is affected by the receptor/Go protein (R/Go) ratio. The binding of guanosine-gamma-[35S]thiotriphosphate ([35S]GTPgammaS) served as the indicator of G protein activation. It was stimulated by empty receptors at high receptor densities, and the dose-response curve was shifted to the left by the agonist carbachol and to the right by the antagonist atropine. At an R/Go ratio of 300:100, the rate of [35S]GTPgammaS binding was the same in the presence or absence of 0. 1 mM carbachol. Alcuronium increased the binding of [35S]GTPgammaS at R/Go ratios of <3:100 and diminished it at R/Go ratios of >10:100, similar to previous observations on intact cells expressing muscarinic receptors at different densities. The apparent biphasicity of alcuronium action indicates that the allosteric modulator has at least two effects on muscarinic receptor/G protein interaction but its mechanistic basis is unclear. The "active state" of muscarinic receptors induced by alcuronium probably is different from that induced by carbachol. Changes in the densities of receptors and Go proteins had little effect on the kinetics of [35S]GTPgammaS binding and on receptor affinity for carbachol, provided the R/Go ratio was kept constant. This suggests that the receptors and G proteins are located in microdomains in which their concentrations remain constant, despite variations in the amounts of lipidic membranes in the system.
Product Description
Alcuronium chloride, European Pharmacopoeia (EP) Reference Standard