Merck
  • Mutagenicity of N-OH-MOCA (4-amino-4'-hydroxylamino-bis-3,3'-dichlorodiphenylmethane) and PBQ (2-phenyl-1,4-benzoquinone) in human lymphoblastoid cells.
Mutagenicity of N-OH-MOCA (4-amino-4'-hydroxylamino-bis-3,3'-dichlorodiphenylmethane) and PBQ (2-phenyl-1,4-benzoquinone) in human lymphoblastoid cells.
Toxicology letters (1998-08-15)
T M Reid, D G DeBord, K L Cheever, R E Savage
ABSTRACT
The genotoxic potential of two occupationally significant chemicals, 4,4'-methylene-bis-2-chloroaniline (MOCA) and 2-phenyl-1,4-benzoquinone (PBQ), was explored by monitoring the induction of mutations at the HPRT locus of AHH-1 human lymphoblastoid cells. Exposure of AHH-1 cells to the putative carcinogenic metabolite of MOCA, N-OH-MOCA, induced a 6-fold increase in mutant frequency and resulted in base pair substitutions primarily at A:T base pairs. In contrast, exposure to PBQ did not result in an increased mutant frequency although this compound was significantly more cytotoxic than N-OH-MOCA at equimolar doses. The induction of mutations at A:T sites by N-OH-MOCA is consistent with the type of DNA damage known to be produced by MOCA and provides a specific marker of genotoxic damage for exposed populations.
Sigma-Aldrich
4,4′-Methylene-bis(2-chloroaniline), 85%