Merck
  • High-pressure liquid chromatographic analysis of pramoxine hydrochloride in high lipoid aerosol foam dosage form.
High-pressure liquid chromatographic analysis of pramoxine hydrochloride in high lipoid aerosol foam dosage form.
Journal of pharmaceutical sciences (1980-04-01)
R Weinberger, B Mann, J Posluszny
ABSTRACT
A rapid and quantitative method for the determination of pramoxine hydrochloride by high-pressure liquid chromatography is presented. The drug is extracted as the salt from a preparation with a high lipoid composition by partitioning it to the aqueous phase of an ether-methanol-water-acetic acid system. The extract is chromatographed on an octadecylsilane bonded packing with a methanol-water-acetic acid-methanesulfonic acid mobile phase. The time required for each separation is approximately 6 min. Analytical recoveries of 100.4 +/- 1.5% were obtained.
Product Description
Supelco
Pramoxine hydrochloride, analytical standard, ≥98%