Merck
  • Catalepsy produced by long-acting neuroleptics, fluphenazine enanthate and fluphenazine decanoate, in mice.
Catalepsy produced by long-acting neuroleptics, fluphenazine enanthate and fluphenazine decanoate, in mice.
Pharmacology (1982-01-01)
K Yamada, T Furukawa
ABSTRACT
The cataleptic state induced by the depot-type preparations of fluphenazine (FZ) was investigated in mice. Intramuscular injections of FZ-enanthate or FZ-decanoate produced a marked and long-lasting catalepsy, which was antagonized by trihexyphenidyl, biperiden and promethazine but not by l-dopa. These anticataleptic effects disappeared after several hours and catalepsy occurred again. The cataleptic effects of FZ-enanthate and FZ-decanoate were increased by (D-Ala2, MePhe4, Met(0)5-ol)-enkephalin, aminooxyacetic acid and haloperidol. The results indicate that the cataleptic effects of FZ-enanthate and FZ-decanoate correspond with their long duration of antipsychotic effects and that the catalepsy may involve enkephalinergic, GABAergic and cholinergic as well as dopaminergic neuron activity.
Product Description
Fluphenazine enantate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.