Merck
  • Hypolipidemic and antioxidant activities of polysaccharides from Rosae Laevigatae Fructus in rats.
Hypolipidemic and antioxidant activities of polysaccharides from Rosae Laevigatae Fructus in rats.
Carbohydrate polymers (2013-04-03)
Chen-Huan Yu, Xiao-Yan Dai, Qin Chen, Jia-Na Zang, Li-Li Deng, Yue-Huan Liu, Hua-Zhong Ying
ABSTRACT
Two major fractions (RLP-1 and RLP-2) were obtained by purifying the crude polysaccharides extracted from a traditional Chinese herb Rosae Laevigatae Fructus. The average molecular weight of RLP-1 and RLP-2 was 21.5 kDa and 16.1 kDa, respectively. Monosaccharide analysis indicated that RLP-1 was composed of xylose, mannose and galactose in the molar ratio of 1:11:8, while RLP-2 was only a glucan. Oral administration of RLP-1 could significantly decrease levels of serum total cholesterol (TC), triglycerides (TG) and low-density lipoprotein cholesterol (LDL-C), inhibit hepatic lipid accumulation, increase antioxidant lipids and up-regulate expressions of peroxisome proliferator-activated receptor-γ (PPAR-γ) and lipoprotein lipase (LPL) in hyperlipidemia rats. These results suggest that RLP-1 improve hyperlipidemia possibly through regulating PPAR-mediated lipid metabolism. Therefore, could be explored as a possible agent for hyperlipidemia.
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Pricing and availability is not currently available.
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.