Merck
  • Serum amyloid P is a sialylated glycoprotein inhibitor of influenza A viruses.
Serum amyloid P is a sialylated glycoprotein inhibitor of influenza A viruses.
PloS one (2013-04-02)
Emma R Job, Barbara Bottazzi, Brad Gilbertson, Kathryn M Edenborough, Lorena E Brown, Alberto Mantovani, Andrew G Brooks, Patrick C Reading
ABSTRACT
Members of the pentraxin family, including PTX3 and serum amyloid P component (SAP), have been reported to play a role in innate host defence against a range of microbial pathogens, yet little is known regarding their antiviral activities. In this study, we demonstrate that human SAP binds to human influenza A virus (IAV) strains and mediates a range of antiviral activities, including inhibition of IAV-induced hemagglutination (HA), neutralization of virus infectivity and inhibition of the enzymatic activity of the viral neuraminidase (NA). Characterization of the anti-IAV activity of SAP after periodate or bacterial sialidase treatment demonstrated that α(2,6)-linked sialic acid residues on the glycosidic moiety of SAP are critical for recognition by the HA of susceptible IAV strains. Other proteins of the innate immune system, namely human surfactant protein A and porcine surfactant protein D, have been reported to express sialylated glycans which facilitate inhibition of particular IAV strains, yet the specific viral determinants for recognition of these inhibitors have not been defined. Herein, we have selected virus mutants in the presence of human SAP and identified specific residues in the receptor-binding pocket of the viral HA which are critical for recognition and therefore susceptibility to the antiviral activities of SAP. Given the widespread expression of α(2,6)-linked sialic acid in the human respiratory tract, we propose that SAP may act as an effective receptor mimic to limit IAV infection of airway epithelial cells.
Sigma-Aldrich
N-Acetylneuraminic acid, ≥95% anhydrous basis, synthetic
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetylneuraminic acid, ≥98% (HPLC), from Escherichia coli
Pricing and availability is not currently available.