Merck
  • Identification of novel imidazo[1,2-a]pyridine inhibitors targeting M. tuberculosis QcrB.
Identification of novel imidazo[1,2-a]pyridine inhibitors targeting M. tuberculosis QcrB.
PloS one (2013-01-10)
Katherine A Abrahams, Jonathan A G Cox, Vickey L Spivey, Nicholas J Loman, Mark J Pallen, Chrystala Constantinidou, Raquel Fernández, Carlos Alemparte, Modesto J Remuiñán, David Barros, Lluis Ballell, Gurdyal S Besra
ABSTRACT
Mycobacterium tuberculosis is a major human pathogen and the causative agent for the pulmonary disease, tuberculosis (TB). Current treatment programs to combat TB are under threat due to the emergence of multi-drug and extensively-drug resistant TB. Through the use of high throughput whole cell screening of an extensive compound library a number of imidazo[1,2-a]pyridine (IP) compounds were obtained as potent lead molecules active against M. tuberculosis and Mycobacterium bovis BCG. The IP inhibitors (1-4) demonstrated minimum inhibitory concentrations (MICs) in the range of 0.03 to 5 µM against a panel of M. tuberculosis strains. M. bovis BCG spontaneous resistant mutants were generated against IP 1, 3, and 4 at 5× MIC and subsequent whole genome sequencing identified a single nucleotide polymorphism (937)ACC>(937)GCC (T313A) in qcrB, which encodes the b subunit of the electron transport ubiquinol cytochrome C reductase. This mutation also conferred cross-resistance against IP 1, 3 and 4 demonstrating a common target. Gene dosage experiments confirmed M. bovis BCG QcrB as the target where over-expression in M. bovis BCG led to an increase in MIC from 0.5 to >8 µM for IP 3. An acute murine model of TB infection established bacteriostatic activity of the IP series, which await further detailed characterization.
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%