Merck
  • The practice of therapeutic hypothermia after cardiac arrest in France: a national survey.
The practice of therapeutic hypothermia after cardiac arrest in France: a national survey.
PloS one (2012-10-11)
Jean-Christophe Orban, Florian Cattet, Jean-Yves Lefrant, Marc Leone, Samir Jaber, Jean-Michel Constantin, Bernard Allaouchiche, Carole Ichai
ABSTRACT
Cardiac arrest is a major health concern worldwide accounting for 375,000 cases per year in Europe with a survival rate of <10%. Therapeutic hypothermia has been shown to improve patients' neurological outcome and is recommended by scientific societies. Despite these guidelines, different surveys report a heterogeneous application of this treatment. The aim of the present study was to evaluate the clinical practice of therapeutic hypothermia in cardiac arrest patients. This self-declarative web based survey was proposed to all registered French adult intensive care units (ICUs) (n=357). Paediatrics and neurosurgery ICUs were excluded. The different questions addressed the structure, the practical modalities of therapeutic hypothermia and the use of prognostic factors in patients admitted after cardiac arrest. One hundred and thirty-two out of 357 ICUs (37%) answered the questionnaire. Adherence to recommendations regarding the targeted temperature and hypothermia duration were 98% and 94% respectively. Both guidelines were followed in 92% ICUs. During therapeutic hypothermia, sedative drugs were given in 99% ICUs, mostly midazolam (77%) and sufentanil (59%). Neuromuscular blocking agents (NMBA) were used in 97% ICUs, mainly cisatracurium (77%). Numerous prognostic factors were used after cardiac arrest such as clinical factors (95%), biomarkers (53%), electroencephalography (78%) and evoked potentials (35%). In France, adherence to recommendations for therapeutic hypothermia after cardiac arrest is higher than those previously reported in other countries. Numerous prognostic factors are widely used even if their reliability remains controversial.
Product Description
Supelco
Sufentanil Citrate solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Sufentanil-D5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard