Merck
  • High-power multichannel PPMgLN-based optical parametric oscillator pumped by a master oscillation power amplification-structured Q-switched fiber laser.
High-power multichannel PPMgLN-based optical parametric oscillator pumped by a master oscillation power amplification-structured Q-switched fiber laser.
Applied optics (2012-10-04)
Tao Chen, Kaihua Wei, Peipei Jiang, Bo Wu, Yonghang Shen
ABSTRACT
We experimentally demonstrated a compact fiber laser-pumped multichannel PPMgLN-based optical parametric oscillator (OPO) generating total OPO output power of 15.8, 15.2, 14.2, 12.9, and 8.8 W with idler output power of 4.7, 4.3, 4.1, 3.3, and 2.1 W at the wavelength of 3.43, 3.63, 3.72, 3.83, and 3.99 μm, respectively. The OPO was pumped by a fully fiberized polarization maintaining (PM) ytterbium-doped pulsed fiber master oscillation power amplifier (MOPA) operating at 1064 nm at a repetition rate of 65 kHz with effective pump power of 28.7 W. The MOPA system was constructed with an acousto-optic Q-switched fiber laser seed and only one stage PM fiber amplifier without any free space components, which makes the pump system compact and stable in the long-term. Comparisons on efficiencies and signal wavelength shifts between different channels showed that the idler absorption was the main factor preventing high average-power OPO operation with long idler wavelength.
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis