- Endogenous release of gamma-aminobutyric acid from the medial preoptic area measured by microdialysis in the anaesthetised rat.
Endogenous release of gamma-aminobutyric acid from the medial preoptic area measured by microdialysis in the anaesthetised rat.
The characteristics of gamma-aminobutyric acid (GABA) release as monitored by microdialysis have been investigated in the chloral hydrate anaesthetised rat. The high outflow of GABA following insertion of the microdialysis probe (membrane 2 mm in length, 0.5 mm in diameter) into the medial preoptic area was found to decline to a stable baseline level after 2 h. After this time, perfusion with a medium containing 100 mM potassium ions evoked a 56-fold increase in GABA outflow. The addition of the calcium channel blocker verapamil (100 microM) to the perfusion medium induced significant 25 and 50% reductions in basal and potassium-stimulated GABA outflow, respectively. In the same animals, verapamil caused an 80% decrease in potassium-stimulated noradrenaline outflow. The glutamic acid decarboxylase inhibitors 3-mercaptopropionic acid and L-allylglycine added to the perfusion medium at a concentration of 10 mM reduced basal GABA release by approximately 50% with different time-courses of action. Ethanolamine-O-sulfate, a GABA-transaminase inhibitor, induced significant increases in basal GABA outflow 90 min after inclusion in the perfusion medium. These results demonstrate that microdialysis is a suitable technique with which to monitor extracellular levels of GABA and provide in vivo data on GABA release and degradation mechanisms.