Merck
  • Localization of endogenous morphine-like compounds in the mouse spinal cord.
Localization of endogenous morphine-like compounds in the mouse spinal cord.
The Journal of comparative neurology (2011-11-22)
Alexis Laux, François Delalande, Jinane Mouheiche, Denise Stuber, Alain Van Dorsselaer, Enrica Bianchi, Erwan Bezard, Pierrick Poisbeau, Yannick Goumon
ABSTRACT
Morphine, codeine, morphine-6-glucuronide, and morphine-3-glucuronide are synthesized de novo in mammalian cells and in the central nervous system. Knowledge on endogenous morphine-like compound distribution in the adult mouse brain has been recently improved, and new hypotheses have been suggested about the potential implications in brain physiology. Endogenous morphine-like compounds have been shown to be synthesized in the spinal cord, but their localization is unknown. Here we describe the distribution of endogenous morphine-like compounds (morphine and/or its glucuronides and/or codeine) in the adult mouse spinal cord using a well-validated antibody. By using different microscopy approaches, we found the presence of morphine, codeine, or morphine glucuronides in γ-aminobutyric acid (GABA)-ergic neurons and astrocytes of the spinal cord. Whereas GABAergic neurons containing endogenous morphine-like compounds were located primarily in the ventral horn, astrocytes that were labeled for morphine-like compounds were found throughout the gray matter and the white matter. Our study demonstrates the possibility that endogenous morphine-like compounds in the central nervous system have other functions beyond their analgesic functions.
Product Description
Supelco
Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-3-β-D-glucuronide solution, 100 μg/mL in methanol: water ( 50:50(v/v)), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-6-β-D-glucuronide solution, 1.0 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®