Merck
  • Detection of 1-benzylpiperazine, 1-(3-trifluoromethylphenyl)-piperazine, and 1-(3-chlorophenyl)-piperazine in 3,4-methylenedioxymethamphetamine-positive urine samples.
Detection of 1-benzylpiperazine, 1-(3-trifluoromethylphenyl)-piperazine, and 1-(3-chlorophenyl)-piperazine in 3,4-methylenedioxymethamphetamine-positive urine samples.
Journal of analytical toxicology (2010-01-01)
Amber J Dickson, Shawn P Vorce, Justin M Holler, Timothy P Lyons
ABSTRACT
Historically, ecstasy tablets contained 3,4-methylenedioxymethamphetamine (MDMA) as the psychoactive component. In recent years, the Drug Enforcement Administration (DEA) and other law enforcement agencies have seized ecstasy tablets that are comprised of psychoactive drugs or drug mixtures other than MDMA. Many jurisdictions have reported the presence of piperazine derivatives including 1-benzylpiperazine (BZP), 1-(3-trifluoromethylphenyl)-piperazine (TFMPP), and 1-(3-chlorophenyl)-piperazine (mCPP) in ecstasy tablets. These piperazine derivatives produce stimulant and psychoactive effects similar to those produced by MDMA, amphetamine, and methamphetamine. In many countries, their use is not controlled, and therefore they have become a legal alternative to MDMA. For this study, a targeted population of 251 MDMA-positive urine samples were analyzed for designer drugs, including the piperazine derivatives. A basic liquid-liquid extraction followed by pentafluoropropionic anhydride (PFPA) derivatization and a full scan (m/z 42-550) gas chromatography-mass spectrometry analysis was used to screen the urine samples for 33 designer drugs. Overall, in 36% of the specimens analyzed, a stimulant or psychoactive compound other than MDMA and 3,4-methylenedioxyamphetamine (MDA) was detected. BZP, TFMPP, and mCPP were detected in 15%, 7%, and 1% of the samples, respectively.
Product Description
Supelco
Benzyl piperazine dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®