Merck
  • Enzymatic synthesis of lacto-N-difucohexaose I which binds to Helicobacter pylori.
Enzymatic synthesis of lacto-N-difucohexaose I which binds to Helicobacter pylori.
Methods in enzymology (2010-09-08)
Tatsuo Miyazaki, Takeshi Sato, Kiyoshi Furukawa, Katsumi Ajisaka
ABSTRACT
Helicobacter pylori is known to bind with sugar chains possessing Lewis b structure. We are trying to combine oligosaccharides containing Lewis b sugar chain to water insoluble polysaccharide through some linker. Lacto-N-difucohexaose I (LNDFH I; Fucalpha1-->2Galbeta1-->3[Fucalpha1-->4]GlcNAcbeta1-->3Galbeta1-->4Glc) fits for that purpose, since it consists of Lewis b tetrasaccharide and lactose whose d-glucose residue can be utilized as a linker. We thus developed a method to synthesize this hexaose enzymatically. First, beta-1,3-N-acetylglucosaminyltransferase (beta-1,3-GnT) was partially purified from bovine blood by an established method. Using this enzyme preparation, d-GlcNAc was attached to the d-galactose residue of lactose with a beta-1,3-linkage to produce lacto-N-triose II at 44% yield. The low yield was thought to be due to contaminating N-acetylglucosaminidase that would have hydrolyzed the product, lacto-N-triose II. Next, d-galactose was attached by transglycosylation using ortho-nitrophenyl beta-d-galactopyranoside as a donor with the aid of recombinant beta-1,3-galactosidase from Bacillus circulans to generate lacto-N-tetraose (LNT) at 22% yield. l-Fucose was then linked to the d-galactose residue of LNT via an alpha-1,2-linkage using recombinant human fucosyltransferase I (FUT1) expressed in a baculovirus system (71% yield). The obtained pentasaccharide was subsequently incubated with GDP-beta-l-fucose and commercial fucosyltransferase III (FUT3) to attach l-fucose to the d-GlcNAc residue of LNT with an alpha-1,4-linkage. After purification with an activated carbon column chromatography, 1.7 mg of LNDFH I was obtained (85% yield). We thus produced LNDFH I over four enzymatic steps with a yield of 6%.