- Functional characterization of ttmM unveils new tautomycin analogs and insight into tautomycin biosynthesis and activity.
Functional characterization of ttmM unveils new tautomycin analogs and insight into tautomycin biosynthesis and activity.
Organic letters (2009-03-14)
Jianhua Ju, Wenli Li, Qiuping Yuan, Noel R Peters, F Michael Hoffmann, Scott R Rajski, Hiroyuki Osada, Ben Shen
PMID19281218
ABSTRACT
The biosynthetic gene cluster for tautomycin (TTM), a potent protein phosphatase (PP) inhibitor has recently been characterized. Inactivation of ttmM, which encodes a putative C3' hydroxylase, afforded mutant SB6005 which accumulated three new 3'-deshydroxy TTM analogs, supporting the function of TtmM and the previously proposed linear pathway for TTM biosynthesis. Bioassays reveal the importance of the C3' OH moiety in PP inhibition and that PP inhibition is not the exclusive mechanism driving TTM-induced cell death.