Merck
A novel class of allosteric modulators of AMPA/Kainate receptors.
Bioorganic & medicinal chemistry letters (2009-01-24)
Giuseppe Cannazza, Krzysztof Jozwiak, Carlo Parenti, Daniela Braghiroli, Marina M Carrozzo, Giulia Puia, Gabriele Losi, Mario Baraldi, Wolfgang Lindner, Irving W Wainer
ABSTRACT
The rapid hydrolysis in vivo of IDRA21 to 2-amino-5-chlorobenzensulfonamide has been demonstrated by microdialysis experiments. The IDRA21 metabolite possess in vitro a biological activity similar to that of IDRA21 itself. Taking 2-amino-5-chlorobenzensulfonamide as lead compound, a novel class of AMPAR positive allosteric modulators has been prepared.
Sigma-Aldrich
IDRA 21, ≥98%