- [The clinical pharmacokinetics of kemantane].
[The clinical pharmacokinetics of kemantane].
Farmakologiia i toksikologiia (1991-01-01)
S S Boĭko, V P Zherdev, N A Kisliak
PMID1860503
ABSTRACT
The pharmacokinetics of a new Soviet-made immunostimulant kemantane, a derivative of adamantine, was studied by gas-liquid chromatography in patients with bronchial pathology. It was found that in the blood of the patients kemantane was not practically detected due to a pronounced effect of the "first pass" through the liver. Kemantane kinetics was evaluated by the kinetics of its major active metabolite. On the basis of the performed studies the scheme of using kemantane in the patients with decreased immunity was recommended.