Merck
[Studies on chemical constituents from rhizomes of Panax notoginseng].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-03-08)
Jiang Zeng, Xiu-ming Cui, Jia-ming Zhou, Zhi-yong Jiang, Xue-mei Zhang, Ji-jun Chen
ABSTRACT
To study the chemical constituents from rhizomes of Panax notoginseng. The constituents were isolated and purified by various chromatogragraphic methods, all compounds were identified on the basis of special analysis and physicochemical characters. Eight compounds were isolated from the 80% alcohol extract of the rhizomes and their structures were identified as ginsenoside Rh4 (I), ginsenoside Rh1 (II), ginsenoside Re (III), notoginsenoside R1 (IV) , ginsenoside Rd (V), ginsenoside Rh1 (VI), notoginsenoside S (VII), notoginsenoside T (VIII). Compound I was isolated from this plant for the first time, Compound VII, VIII are isolated from rhizomes of Panax notoginseng for the first time.
Product Description
Supelco
Ginsenoside Rh1, analytical standard
Sigma-Aldrich
Notoginsenoside R1, ≥98% (HPLC)