Merck
  • Novel synthons from mucochloric acid: the first use of alpha,beta-dichloro-gamma-butenolides and gamma-butyrolactams for direct vinylogous aldol addition.
Novel synthons from mucochloric acid: the first use of alpha,beta-dichloro-gamma-butenolides and gamma-butyrolactams for direct vinylogous aldol addition.
The Journal of organic chemistry (2007-04-03)
Koushik Das Sarma, Ji Zhang, Timothy T Curran
ABSTRACT
Novel 5-(1'-hydroxy)-gamma-butyrolactone and gamma-butyrolactam subunits were synthesized by direct vinylogous aldol addition of alpha,beta-dichloro gamma-butyrolactones and gamma-butyrolactams with aldehydes under basic conditions. Different bases and solvents were screened in the context of generating gamma-butyrolactones. Diastereoselectivity was observed and gamma-butenolides and gamma-butyrolactams showed opposite diastereoselectivity under the same reaction condition.
Sigma-Aldrich
Mucochloric acid, 99%