Merck
  • Granular activated carbon adsorption and microwave regeneration for the treatment of 2,4,5-trichlorobiphenyl in simulated soil-washing solution.
Granular activated carbon adsorption and microwave regeneration for the treatment of 2,4,5-trichlorobiphenyl in simulated soil-washing solution.
Journal of hazardous materials (2007-03-21)
Xitao Liu, Gang Yu, Wenya Han
ABSTRACT
The treatment of 2,4,5-trichlorobiphenyl (PCB29) in simulated soil-washing solution by granular activated carbon (GAC) adsorption and microwave (MW) regeneration was investigated in this study. The PCB29 adsorption process was carried out in a continuous flow adsorption column. After adsorption, the PCB29-loaded GAC was dried at 103 degrees C, and regenerated in a quartz reactor by 2450MHz MW irradiation at 700W for 5min. The efficacy of this procedure was analyzed by determining the rates and amounts of PCB29 adsorbed in successive adsorption/MW regeneration cycles. Effects of the regeneration on the textural properties and the PCB29 adsorption capacity of GAC were examined. It was found that after several adsorption/MW regeneration cycles, the adsorption rate of GAC increased, whereas, the adsorption capacity decreased, which could be explained according to the change of textural properties. Most of the PCB29 adsorbed on GAC was degraded within 3min under MW irradiation, and the analysis of degradation products by GC-MS demonstrated that PCB29 experienced dechlorination during this treatment.