Merck
  • TGF-beta1 and integrin synergistically facilitate the differentiation of rat podocytes by increasing alpha-smooth muscle actin expression.
TGF-beta1 and integrin synergistically facilitate the differentiation of rat podocytes by increasing alpha-smooth muscle actin expression.
Translational research : the journal of laboratory and clinical medicine (2006-08-30)
Chien-An Chen, Jyh-Chang Hwang, Jinn-Yuh Guh, Jer-Chia Tsai, Hung-Chun Chen
ABSTRACT
Phenotypic changes can be found in certain glomerular diseases, and the cell origin is not defined. This study was designed to identify whether podocytes can differentiate by the expression of alpha-smooth muscle actin (alpha-SMA), under the effects of TGF-beta(1) (transforming growth factor-beta(1)) and integrin. Western and Northern blot analyses were performed to identify the protein and mRNA (messenger ribonucleic acid) expression of alpha-SMA. The number of podocytes, which express alpha-SMA, was measured by immunocytochemical staining. The results showed that TGF-beta(1) dose-dependently increased alpha-SMA protein and mRNA expression at 4 and 2 days, respectively. TGF-beta(1) also dose-dependently increased the alpha-SMA staining of podocytes. The alpha-SMA-positive podocytes showed front-end and back-end polarity. The integrinalpha3beta(1) antagonists, anti-integrinbeta(1) monoclonal antibody and Gly-Arg-Gly-Asp (GRGD), decreased the expression of alpha-SMA protein and the percentage of alpha-SMA positive cells stimulated by TGF-beta(1) (both P < 0.01). The addition of calphostin [inhibitor of protein kinase C (PKC)] and genistein [inhibitor of focal adhesion kinase (FAK)] also decreased the expression of alpha-SMA protein and the percentage of alpha-SMA positive cells stimulated by TGF-beta(1) (both P < 0.01). In conclusion, this study indicated that TGF-beta(1) may act synergistically with integrins, through activation of PKC and FAK, to induce the phenotypic changes of rat podocytes with increasing alpha-SMA expression.