Merck
  • The effect of alkaline phosphatase inhibitors on intracellular lipid accumulation in preadipocytes isolated from human mammary tissue.
The effect of alkaline phosphatase inhibitors on intracellular lipid accumulation in preadipocytes isolated from human mammary tissue.
Annals of clinical biochemistry (2006-05-18)
Aus T Ali, Clem B Penny, Janice E Paiker, George Psaras, Faisel Ikram, Nigel J Crowther
ABSTRACT
A previous study has demonstrated that alkaline phosphatase (AP) may play a role in the control of intracellular lipid accumulation in the rodent preadipocyte cell line, 3T3-L1. The present study investigated whether AP may have a similar function in preadipocytes isolated from human mammary gland tissue. Preadipocyte maturation was induced in the presence or absence of the tissue non-specific AP inhibitors levamisole and histidine, and the tissue-specific AP inhibitor PheGlyGly. Cellular AP activity and adipogenesis were both assessed at 0 and 12 days post-induction of differentiation. After differentiation, AP activity increased 5.1 +/- 1.3-fold in the absence and 8.9 +/- 2.8-fold (P < 0.05) in the presence of levamisole. However, adipogenesis increased 1.95 +/- 0.11-fold in the absence but only 1.36 +/- 0.06-fold (P < 0.001) in the presence of levamisole. There was a 4.2 +/- 2.2-fold increase in AP activity in the absence and a 0.51 +/- 0.46-fold (P < 0.05) decrease in the presence of histidine. Adipogenesis increased 2.09 +/- 0.35-fold in the absence of histidine but only 1.22 +/- 0.30-fold (P < 0.05) in the presence of histidine. PheGlyGly had no effects. Fluorescent microscopy showed AP activity was localized to the triglyceride-containing droplets of the cell. This is the first study to show that tissue non-specific AP inhibitors can block adipogenesis in human preadipocytes.
Sigma-Aldrich
Phe-Gly-Gly