Merck
The simple SDD.
Radiation protection dosimetry (2006-04-29)
A C Fernandes, T Morlat, M Felizardo, J I Collar, J Puibasset, G Waysand, H S Miley, A R Ramos, T A Girard, F Giuliani, D Limagne, J G Marques, R C Martins, C Oliveira
ABSTRACT
We describe the fabrication and characterisation of the SIMPLE superheated droplet detector, a 10 g active mass device of C(2)ClF(5) in 1-3% weight concentrations currently employed in a direct search for spin-dependent astroparticle dark matter candidates.
Sigma-Aldrich
Chloropentafluoroethane, ≥98%