Merck
  • Anthranilic acid based CCK1 receptor antagonists and CCK-8 have a common step in their "receptor desmodynamic processes".
Anthranilic acid based CCK1 receptor antagonists and CCK-8 have a common step in their "receptor desmodynamic processes".
Journal of medicinal chemistry (2006-04-14)
Stefania De Luca, Michele Saviano, Lucia Lassiani, Konstantina Yannakopoulou, Penny Stefanidou, Luigi Aloj, Giancarlo Morelli, Antonio Varnavas
ABSTRACT
The interaction between the 1-47 N-terminus of the CCK(1)-R and the anthranilic acid based antagonists has been investigated by fluorescence spectroscopy. These antagonists interact with W39 of the N-terminal domain of the CCK(1)-R like that of the endogenous ligand CCK-8. This specific interaction was not found in other nonpeptide ligands of the CCK(1)-R. Conformational studies, using NMR and energy minimization procedures, have allowed formulation of a new hypothesis on the CCK(1)-R binding mode of the anthranilic antagonists.
Sigma-Aldrich
Loxiglumide, ≥97% (HPLC)