Merck
  • Supercritical water oxidation of the PCB congener 2-chlorobiphenyl in methanol solutions: a kinetic analysis.
Supercritical water oxidation of the PCB congener 2-chlorobiphenyl in methanol solutions: a kinetic analysis.
Environmental science & technology (2005-09-30)
Christopher P O'Brien, Mark C Thies, David A Bruce
ABSTRACT
Incineration is commonly used to destroy polychlorinated biphenyl (PCB) wastes, but this method of treatment is not ideal for all mixed liquid wastes, especially those containing radioactive materials. Therefore, other remediation technologies are needed to efficiently treat these waste forms. This study examined the supercritical water oxidation (SCWO) of 2-chlorobiphenyl (2-PCB), using hydrogen peroxide as the oxidant and methanol as a cosolvent at 2 vol %. Kinetic studies were carried out in a plug flow reactor at temperatures from 686 to 789 K and a pressure of 250 bar, with reactor residence times ranging from 1.1 to 5.8 s. Least-squares regression of the collective reaction rate data revealed that 2-PCB exhibited second-order kinetics, with an Arrhenius frequency factor, A, equal to 10(18.2+/-2.3) L x mol(-1) x s(-1) and activation energy, Ea, of 181.7 +/- 33.2 kJ/mol. The primary organic reaction products from the SCWO of 2-PCB were biphenyl at low temperatures (<700 K) and CO2 at elevated temperatures. No dioxins or chlorinated dibenzofurans were ever detected in any of the effluent samples. Additional experiments with higher organic feed concentrations (4 vol %) illustrated how the exothermic nature of the organic oxidation reactions can be used to render the process self-sustaining. Finally, SCWO destruction rates greater than 99.98% were achieved for a simulated PCB-contaminated job control waste similar to that encountered at many U.S. Department of Energy sites.