- Groundwater contamination by chlorinated naphthalenes and related substances caused by activities of a former military base.
Groundwater contamination by chlorinated naphthalenes and related substances caused by activities of a former military base.
Water samples derived from two different aquifer layers of six sampling sites were analysed by GC/MS in order to characterize a groundwater contamination caused by chemicals used for wood impregnation. Mono- and dichlorinated naphthalenes, chlorobenzo(b)thiophene, 1-chloro-4-naphthol, 1-chloronaphthoic acid, acenaphthene and methyled naphthalenes were identified as the main pollutants and quantified. 1-Chloro-4-naphthol and 1-chloronaphthoic acid are discussed as possible indicators for anaerobic degradation processes. Results of inorganic and compound specific stable carbon isotope analyses revealed only a minor degree of microbiological transformation. Thus, sorption was characterized as the main attenuation process within the aquifer affecting the contamination described.