Merck
Enzymatic synthesis of D-glucosaminic acid from D-glucosamine.
Carbohydrate research (2004-12-29)
Fabio Pezzotti, Hélène Therisod, Michel Therisod
ABSTRACT
D-glucosaminic acid (2-amino-2-deoxy-D-gluconic acid), a component of bacterial lipopolysaccharides and a chiral synthon, is easily prepared on a multigram scale by air oxidation of D-glucosamine (2-amino-2-deoxy-D-glucose) catalysed by glucose oxidase.
Sigma-Aldrich
D-Glucosaminic acid, ≥98% (TLC)