Merck
  • Effects of the acyl coenzyme A:cholesterol acyltransferase inhibitor avasimibe on human atherosclerotic lesions.
Effects of the acyl coenzyme A:cholesterol acyltransferase inhibitor avasimibe on human atherosclerotic lesions.
Circulation (2004-11-10)
Jean-Claude Tardif, Jean Grégoire, Philippe L L'Allier, Todd J Anderson, Olivier Bertrand, Francois Reeves, Lawrence M Title, Fernando Alfonso, Erick Schampaert, Alita Hassan, Richard McLain, Milton L Pressler, Reda Ibrahim, Jacques Lespérance, John Blue, Therese Heinonen, Josep Rodés-Cabau
ABSTRACT
Inhibition of the acyl coenzyme A:cholesterol acyltransferase (ACAT) enzyme may prevent excess accumulation of cholesteryl esters in macrophages. The ACAT inhibitor avasimibe was shown to reduce experimental atherosclerosis. This study was designed to investigate the effects of avasimibe on human coronary atherosclerosis. This randomized, double-blind, placebo-controlled trial assessed the effects of avasimibe at dosages of 50, 250, and 750 mg QD on the progression of coronary atherosclerosis as assessed by intravascular ultrasound (IVUS). All patients received background lipid-lowering therapy if necessary to reach a target baseline LDL level <125 mg/dL (3.2 mmol/L). IVUS and coronary angiography were performed at baseline and repeated after up to 24 months of treatment. Approximately equal percentages of patients across groups received concurrent statin therapy (87% to 89%). The mean total plaque volume at baseline was approximately 200 mm3, and the least squares mean change at end of treatment was 0.7 mm3 for placebo and 7.7, 4.1, and 4.8 mm3 for the avasimibe 50, 250, and 750 mg groups, respectively (adjusted P=0.17 [unadjusted P=0.057], 0.37, and 0.37, respectively). Percent atheroma volume increased by 0.4% with placebo and by 0.7%, 0.8%, and 1.0% in the respective avasimibe groups (P=NS). LDL cholesterol increased during the study by 1.7% with placebo but by 7.8%, 9.1%, and 10.9% in the respective avasimibe groups (P<0.05 in all groups). Avasimibe did not favorably alter coronary atherosclerosis as assessed by IVUS. This ACAT inhibitor also caused a mild increase in LDL cholesterol.
Sigma-Aldrich
Avasimibe, ≥98% (HPLC)