Merck
  • Chemoenzymatic synthesis of optically active, biodegradable polymers based on phenyl- and naphthyl-ethanols esterified with divinyladipate.
Chemoenzymatic synthesis of optically active, biodegradable polymers based on phenyl- and naphthyl-ethanols esterified with divinyladipate.
Biotechnology letters (2004-06-18)
Takao Raku, Yutaka Tokiwa
ABSTRACT
For the purpose of developing a new synthetic polymer containing an asymmetric molecule branch, three racemic alcohols, i.e. 1-phenylethanol, 1-(4-methylphenyl)ethanol and 1-(2-naphthyl)ethanol, were esterified enzymatically with divinyladipate using a lipase from Pseudomonas cepacia. The enzymatic acylation of alcohols produced monoacylated products. Optically active polymerizable monomers, (R)-vinyl adipic acid (phenyl-1-yl) ethyl ester, (R)-vinyl adipic acid (4-methylphenyl-1-yl) ethyl ester and (R)-vinyl adipic acid (2-naphthyl-1-yl) ethyl ester with enantiometric excesses over 99%, 96% and 99%, respectively, were obtained. Each optically active monomer was then subjected to free radical polymerization, to give polymers having a number average molecular weight of 2.9 x 10(3) - 2.2 x 10(4). These polymers are considered useful as optically active polymers having biodegradability.
Sigma-Aldrich
2-Methyl-1-phenyl-2-propanol, 98%