Merck
HPLC analysis of hexosamine phosphates in biological samples.
Journal of biochemical and biophysical methods (1992-12-01)
S Ambrosio, J R De Oliveira, R Bartrons
ABSTRACT
Galactosamine is quickly metabolized to galactosamine 1-phosphate in rats treated with this compound. An HPLC method to quantify hexosamine phosphates in biological samples is described, modified from the o-phthaldialdehyde amino acid analysis procedure. o-Phthaldialdehyde derivatives of hexosamines and hexosamine-phosphates can be eluted from a reverse-phase column at different retention times, with a total analysis time of 30 min and without overlapping with free amino acids at physiological concentrations. The standard curves are linear between 1 and 40 nmol. This simple method is more selective and sensitive than previous enzymatic analyses of hexosamine phosphorylation.
Sigma-Aldrich
α-D-Glucosamine 1-phosphate