Merck
  • Glutamate-mediated cytosolic calcium oscillations regulate a pulsatile prostaglandin release from cultured rat astrocytes.
Glutamate-mediated cytosolic calcium oscillations regulate a pulsatile prostaglandin release from cultured rat astrocytes.
The Journal of physiology (2003-09-23)
Micaela Zonta, Annalisa Sebelin, Sara Gobbo, Tommaso Fellin, Tullio Pozzan, Giorgio Carmignoto
ABSTRACT
The synaptic release of glutamate evokes in astrocytes periodic increases in [Ca2+]i, due to the activation of metabotropic glutamate receptors (mGluRs). The frequency of these [Ca2+]i oscillations is controlled by the level of neuronal activity, indicating that they represent a specific, frequency-coded signalling system of neuron-to-astrocyte communication. We recently found that neuronal activity-dependent [Ca2+]i oscillations in astrocytes are the main signal that regulates the coupling between neuronal activity and blood flow, the so-called functional hyperaemia. Prostaglandins play a major role in this fundamental phenomenon in brain function, but little is known about a possible link between [Ca2+]i oscillations and prostaglandin release from astrocytes. To investigate whether [Ca2+]i oscillations regulate the release of vasoactive prostaglandins, such as the potent vasodilator prostaglandin E2 (PGE2), from astrocytes, we plated wild-type human embryonic kidney (HEK)293 cells, which respond constitutively to PGE2 with [Ca2+]i elevations, onto cultured astrocytes, and used them as biosensors of prostaglandin release. After loading the astrocyte-HEK cell co-cultures with the calcium indicator Indo-1, confocal microscopy revealed that mGluR-mediated [Ca2+]i oscillations triggered spatially and temporally coordinated [Ca2+]i increases in the sensor cells. This response was absent in a clone of HEK cells that are unresponsive to PGE2, and recovered after transfection with the InsP3-linked prostanoid receptor EP1. We conclude that [Ca2+]i oscillations in astrocytes regulate prostaglandin releases that retain the oscillatory behaviour of the [Ca2+]i changes. This finely tuned release of PGE2 from astrocytes provides a coherent mechanistic background for the role of these glial cells in functional hyperaemia.
Sigma-Aldrich
SC 19220, ≥98% (HPLC), solid