Merck
  • Interventions for smoking cessation and reduction in individuals with schizophrenia.
Interventions for smoking cessation and reduction in individuals with schizophrenia.
The Cochrane database of systematic reviews (2013-03-02)
Daniel T Tsoi, Mamta Porwal, Angela C Webster
ABSTRACT
Individuals with schizophrenia smoke more heavily than the general population and this contributes to their higher morbidity and mortality from smoking-related illnesses. It remains unclear what interventions can help them to quit or to reduce smoking. To evaluate the benefits and harms of different treatments for nicotine dependence in schizophrenia. We searched electronic databases including MEDLINE, EMBASE and PsycINFO from inception to October 2012, and the Cochrane Tobacco Addiction Group Specialized Register in November 2012. We included randomised trials for smoking cessation or reduction, comparing any pharmacological or non-pharmacological intervention with placebo or with another therapeutic control in adult smokers with schizophrenia or schizoaffective disorder. Two reviewers independently assessed the eligibility and quality of trials, as well as extracted data. Outcome measures included smoking abstinence, reduction in the amount smoked and any change in mental state. We extracted abstinence and reduction data at the end of treatment and at least six months after the intervention. We used the most rigorous definition of abstinence or reduction and biochemically validated data where available. We noted any reported adverse events. Where appropriate, we pooled data using a random-effects model. We included 34 trials (16 trials of cessation; nine trials of reduction; one trial of relapse prevention; eight trials that reported smoking outcomes for interventions aimed at other purposes). Seven trials compared bupropion with placebo; meta-analysis showed that cessation rates after bupropion were significantly higher than placebo at the end of treatment (seven trials, N = 340; risk ratio [RR] 3.03; 95% confidence interval [CI] 1.69 to 5.42) and after six months (five trials, N = 214, RR 2.78; 95% CI 1.02 to 7.58). There were no significant differences in positive, negative and depressive symptoms between bupropion and placebo groups. There were no reports of major adverse events such as seizures with bupropion.Smoking cessation rates after varenicline were significantly higher than placebo, at the end of treatment (2 trials, N = 137; RR 4.74, 95% CI 1.34 to 16.71). Only one trial reported follow-up at six months and the CIs were too wide to provide evidence of a sustained effect (one trial, N = 128, RR 5.06, 95% CI 0.67 to 38.24). There were no significant differences in psychiatric symptoms between the varenicline and placebo groups. Nevertheless, there were reports of suicidal ideation and behaviours from two people on varenicline.Two studies reported that contingent reinforcement (CR) with money may increase smoking abstinence rates and reduce the level of smoking in patients with schizophrenia. However, it is uncertain whether these benefits can be maintained in the longer term. There was no evidence of benefit for the few trials of other pharmacological therapies (including nicotine replacement therapy (NRT)) and psychosocial interventions in helping smokers with schizophrenia to quit or reduce smoking. Bupropion increases smoking abstinence rates in smokers with schizophrenia, without jeopardizing their mental state. Varenicline may also improve smoking cessation rates in schizophrenia, but its possible psychiatric adverse effects cannot be ruled out. CR may help this group of patients to quit and reduce smoking in the short term. We failed to find convincing evidence that other interventions have a beneficial effect on smoking in schizophrenia.
Sigma-Aldrich
Bupropion hydrochloride, ≥98% (HPLC), solid
Supelco
Bupropion hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®