Merck
  • Real refractive indices and formation yields of secondary organic aerosol generated from photooxidation of limonene and α-pinene: the effect of the HC/NO(x) ratio.
Real refractive indices and formation yields of secondary organic aerosol generated from photooxidation of limonene and α-pinene: the effect of the HC/NO(x) ratio.
The journal of physical chemistry. A (2012-04-19)
Hwajin Kim, Brian Barkey, Suzanne E Paulson
ABSTRACT
The refractive index is an important property affecting aerosol optical properties, which in turn help determine the aerosol direct effect and satellite retrieval results. Here, we investigate the real refractive indices (m(r)) of secondary organic aerosols (SOA) generated from the photooxidation of limonene and α-pinene with different HC/NO(x) ratios. Refractive indices were obtained from polar nephelometer data using parallel and perpendicular polarized 532 nm light combined with measured size distributions, and retrievals were performed using a genetic algorithm and Mie-Lorenz scattering theory. The absolute error associated with the m(r) retrieval is ±0.03, and reliable retrievals are possible for mass concentrations above 5-20 μg/m(3) depending on particle size. The limonene SOA data suggest the most important factor controlling the refractive index is the HC/NO(x) ratio; the refractive index is much less sensitive to the aerosol age or mass concentration. The refractive index ranges from about 1.34 to 1.56 for limonene and from 1.36 to 1.52 for α-pinene, and generally decreases as the HC/NO(x) ratio increases. Especially for limonene, the particle diameter is also inversely related to the HC/NO(x) ratio; the final size mode increases from 220 to 330 nm as the HC/NO(x) ratio decreases from 33 to 6. In an effort to explore the ability of models from the literature to explain the observed refractive indices, a recent limonene oxidation mechanism was combined with SOA partitioning and a structure-property relationship for estimating refractive indices of condensing species. The resulting refractive indices fell in a much narrower range (1.475 ± 0.02) of m(r) than observed experimentally. We hypothesize the experimentally observed high m(r) values are due to oligomerization and the low values to water uptake, small soluble molecules such as glyoxal and other factors, each of which is not included in the oxidation mechanism. Aerosol formation yields were measured over the mass concentration range from 6 to ∼150 μg/m(3), over which they increased steadily, and were higher for high HC/NO(x) ratio experiments.
Product Description
α-Pinene, primary reference standard
Sigma-Aldrich
α-Pinene, 98%