Merck
  • Limonin methoxylation influences the induction of glutathione S-transferase and quinone reductase.
Limonin methoxylation influences the induction of glutathione S-transferase and quinone reductase.
Journal of agricultural and food chemistry (2009-06-02)
Jose Luis Perez, G K Jayaprakasha, Violeta Valdivia, Diana Munoz, Deepak V Dandekar, Hassan Ahmad, Bhimanagouda S Patil
ABSTRACT
Previous studies have indicated the chemopreventive potential of citrus limonoids due to the induction of phase II detoxifying enzymes. In the present study, three citrus limonoids were purified and identified from sour orange seeds as limonin, limonin glucoside (LG), and deacetylnomilinic acid glucoside (DNAG). In addition, limonin was modified to defuran limonin and limonin 7-methoxime. The structures of these compounds were confirmed by NMR studies. These five compounds were used to investigate the influence of phase II enzymes in female A/J mice. Our results indicated the highest induction of glutathione S-transferase (GST) activity against 1-chloro-2,4-dinitrobenzene (CDNB) by DNAG (67%) in lung homogenates followed by limonin-7-methoxime (32%) in treated liver homogenates. Interestingly, limonin-7-methoxime showed the highest GST activity (270%) in liver against 4-nitroquinoline 1-oxide (4NQO), while the same compound in the stomach induced GST by 51% compared to the control. The DNAG treated group induced 55% in stomach homogenates. Another phase II enzyme, quinone reductase (QR), was significantly induced by limonin-7-methoxime by 65 and 32% in liver and lung homogenates, respectively. Defuran limonin induced QR in lung homogenates by 45%. Our results indicated that modification of limonin has differential induction of phase II enzymes. These findings are indicative of a possible mechanism for the prevention of cancer by aiding in the detoxification of xenobiotics.
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)