Merck
  • Repulsion between inorganic particles inserted within surfactant bilayers.
Repulsion between inorganic particles inserted within surfactant bilayers.
Physical review letters (2008-10-15)
Doru Constantin, Brigitte Pansu, Marianne Impéror, Patrick Davidson, François Ribot
ABSTRACT
We study by synchrotron small-angle x-ray scattering highly aligned lamellar phases of a zwitterionic surfactant, doped with monodisperse and spherical hydrophobic inorganic particles as a function of particle concentration. Analysis of the structure factor of the two-dimensional fluid formed by the particles in the plane of the bilayer gives access to their membrane-mediated interaction, which is repulsive, with a contact value of about 4kBT and a range of 14 angstroms. Systematic application of this technique should lead to a better understanding of the interaction between membrane inclusions.
Sigma-Aldrich
N,N-Dimethyldodecylamine N-oxide, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
N,N-Dimethyldodecylamine N-oxide solution, ~30% in H2O