Merck
Effect of 3-hydroxyanthranilic acid on mushroom tyrosinase activity.
Biochimica et biophysica acta (1998-07-11)
A Rescigno, E Sanjust, G Soddu, A C Rinaldi, F Sollai, N Curreli, A Rinaldi
ABSTRACT
Tyrosinase is a copper containing protein which catalyzes the hydroxylation of monophenols and the oxidation of diphenols to o-quinones. The monophenolase activity of tyrosinase is characterized by a typical lag time. In this paper the influence of 3-hydroxyanthranilic acid on monophenolase activity of tyrosinase is reported. 3-Hydroxyanthranilic acid reduced the lag time of tyrosinase when the enzyme acted on N-acetyl-L-tyrosine and on 4-tert-butylphenol. In the presence of 3-hydroxyanthranilic acid, the reaction product 4-tert-butyl-o-benzoquinone, derived from 4-tert-butylphenol oxidation, was formed at a higher rate than in its absence. The results reported in this paper indicate that 3-hydroxyanthranilic acid could affect the enzymic activity of mushroom tyrosinase probably by acting as a diphenol substrate. A K(m) value of 0.78 mM was calculated for 3-hydroxyanthranilic acid as substrate. When tyrosinase acted on 4-tert-butylphenol, K(m) for 3-hydroxyanthranilic acid as a cofactor was estimated to be 37.5 microM. No effect was observed on the diphenolase activity of the enzyme acting on 4-tert-butylcatechol in the presence of 3-hydroxyanthranilic acid.
Product Description
Supelco
N-Acetyl-L-tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material