Merck
  • A separation of clinical from epidermal thinning effect in the topical glucocorticoid clobetasone butyrate.
A separation of clinical from epidermal thinning effect in the topical glucocorticoid clobetasone butyrate.
The British journal of dermatology (1977-01-01)
D V Stevanovic, L Wilson, C G Sparkes
PMID320995
ABSTRACT
Clobetasone butyrate ointment has been shown to be more effective in treating psoriasis and eczema than flurandrenolone ointment yet to cause less epidermal thinning in a human experimental model. This is an indication that the clinical activity of topical glucocorticoids may not necessarily be inseparable from their propensity to cause atrophy of the skin.